missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxyindole-2-carboxylate, 99%

Product Code. 15453767
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453767 250 mg 250mg
11362889 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453767 Supplier Thermo Scientific Alfa Aesar Supplier No. H27743.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111258-23-2 | C11H11NO3 | 205.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111258-23-2
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD00134300
InChI Key GLCZQTLCVLVFGV-UHFFFAOYSA-N
Synonym methyl 4-methoxy-2-indolecarboxylate, 4-methoxy-1h-indole-2-carboxylic acid methyl ester, methyl 4-methoxyindole-2-carboxylate, 1h-indole-2-carboxylic acid, 4-methoxy-, methyl ester, 4-methoxyindole-2-carboxylic acid methyl ester, methyl-4-methoxy-2-indolecarboxylate, 1h-indole-2-carboxylicacid, 4-methoxy-, methyl ester, methyl4-methoxy-2-indolecarboxylate, pubchem7250, acmc-1btz2
PubChem CID 688172
IUPAC Name methyl 4-methoxy-1H-indole-2-carboxylate
SMILES COC(=O)C1=CC2=C(OC)C=CC=C2N1

Specifications

Melting Point 140°C to 142°C
Quantity 250 mg
Formula Weight 205.21
Percent Purity 99%
Chemical Name or Material Methyl 4-methoxyindole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.