missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methoxybenzoate, Mikromol™

Product Code. 17939404
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17939404

Brand: Mikromol MM1063.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 4-Methoxybenzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 4-Methoxybenzoate
CAS 121-98-2
Analyte or Component Names Methyl 4-Methoxybenzoate
Shipping Condition Room Temperature
Molecular Formula C9 H10 O3
Certifications/Compliance ISO 17025
Synonym p-Anisic acid, methyl ester (6CI,7CI,8CI), 4-(Methoxycarbonyl)anisole, 4-Methoxybenzoic acid methyl ester, Methyl 4-methoxybenzoate, Methyl p-anisate, Methyl p-methoxybenzoate, NSC 7324, p-Methoxybenzoic acid methyl ester, Methyl Salicylate Impurity D
InChI Formula InChI=1S/C9H10O3/c1-11-8-5-3-7(4-6-8)9(10)12-2/h3-6H,1-2H3
SMILES COC(=O)c1ccc(OC)cc1
IUPAC Name methyl 4-methoxybenzoate
Molecular Weight (g/mol) 166.17
Quantity 100 mg
Formula Weight 166.063 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.