missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxybenzoate, 98+%

Product Code. p-7020588
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11434324

Brand: Thermo Scientific Alfa Aesar A12598.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-98-2 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-98-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008437
InChI Key DDIZAANNODHTRB-UHFFFAOYSA-N
Synonym methyl anisate, methyl p-anisate, methyl p-methoxybenzoate, 4-methoxybenzoic acid methyl ester, p-anisic acid, methyl ester, benzoic acid, 4-methoxy-, methyl ester, methyl 4-anisate, p-methoxybenzoic acid methyl ester, p-anisic acid methyl ester, benzoic acid, p-methoxy-, methyl ester
PubChem CID 8499
ChEBI CHEBI:86903
IUPAC Name methyl 4-methoxybenzoate
SMILES COC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 48°C to 51°C
Boiling Point 244°C to 245°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
Beilstein 2208571
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material Methyl 4-methoxybenzoate
compliance-icons
Product Identifier
  • Methyl 4-methoxybenzoate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 4-methoxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.