missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodobutyrate, 95%, stabilized

Product Code. 10294614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10294614 5 g Glass bottle 5g
10493074 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10294614

Brand: Thermo Scientific Acros 426370050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14273-85-9 | C5H9IO2 | 228.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14273-85-9
Molecular Formula C5H9IO2
Molecular Weight (g/mol) 228.02
MDL Number MFCD00674066
InChI Key NBCIIVXSBPDKOM-UHFFFAOYSA-N
Synonym methyl 4-iodobutyrate, butanoic acid, 4-iodo-, methyl ester, butyric acid, 4-iodo-, methyl ester, methyl-4-iodobutyrate, acmc-1bwlv, methyl gamma-iodobutyrate, 4-iodobutyric acid methyl ester, butanoic acid, 4-iodo-,methyl ester
PubChem CID 547991
IUPAC Name methyl 4-iodobutanoate
SMILES COC(=O)CCCI

Specifications

Color Colorless to Yellow
Density 1.6890g/mL
Boiling Point 80°C to 83°C (11.0 mmHg)
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula I(CH2)3COOCH3
Quantity 5 g
Specific Gravity 1.689
Stabilizer Copper
Formula Weight 228.02
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 4-iodobutyrate, Stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.