missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodobenzoate, 98%

Product Code. 11453947
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453947 250 g 250g
11433947 10 g 10g
11443947 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453947 Supplier Thermo Scientific Alfa Aesar Supplier No. A19342.30
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 619-44-3 | C8H7IO2 | 262.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-44-3
Molecular Formula C8H7IO2
MDL Number MFCD00016353
InChI Key DYUWQWMXZHDZOR-UHFFFAOYSA-N
Synonym methyl p-iodobenzoate, 4-iodobenzoic acid methyl ester, benzoic acid, 4-iodo-, methyl ester, 4-iodo-benzoic acid methyl ester, benzoic acid, p-iodo-, methyl ester, methyl-4-iodobenzoate, methyl 4-iodo-benzoate, methyl-4-iodo-benzoate, pubchem20378, methyl para-iodobenzoate
PubChem CID 69273
IUPAC Name methyl 4-iodobenzoate

Specifications

Melting Point 114°C to 116°C
Quantity 250 g
Sensitivity Light sensitive
SMILES COC(=O)C1=CC=C(I)C=C1
Molecular Weight (g/mol) 262.05
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material Methyl 4-iodobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.