missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodo-3-nitrobenzoate, 99%

Product Code. 11447226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447226 5 g 5g
11457226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447226 Supplier Thermo Scientific Alfa Aesar Supplier No. B25544.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89976-27-2 | C8H6INO4 | 307.043 g/mol

Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Soluble in DCM 10mg/0.5mL. Insoluble in water.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89976-27-2
Molecular Formula C8H6INO4
Molecular Weight (g/mol) 307.043
MDL Number MFCD08064223
InChI Key SCMBIQRYVKITCY-UHFFFAOYSA-N
PubChem CID 297884
IUPAC Name methyl 4-iodo-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)I)[N+](=O)[O-]

Specifications

Melting Point 101°C to 104°C
Quantity 5 g
Sensitivity Light sensitive
Synonym 4-iodo-3-nitrobenzoic acid methyl ester, benzoic acid,4-iodo-3-nitro-, methyl ester, benzoic acid, 4-iodo-3-nitro-, methyl ester, methyl4-iodo-3-nitrobenzoate, acmc-20af7x, methyl-4-iodo-3-nitrobenzoate, 4-iodo-3-nitro benzoic methyl ester, 3-nitro-4-iodobenzoic acid methyl ester, 4-iodo-3-nitro-benzoic acid methyl ester
Solubility Information Soluble in DCM 10mg/0.5mL. Insoluble in water.
Formula Weight 307.05
Percent Purity 99%
Chemical Name or Material Methyl 4-iodo-3-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.