missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodo-3-methylbenzoate, 98%

Product Code. 11431020
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431020 5 g 5g
11441020 25 g 25g
11451020 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431020 Supplier Thermo Scientific Alfa Aesar Supplier No. B25395.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5471-81-8 | C9H9IO2 | 276.073 g/mol

Methyl 4-iodo-3-methylbenzoate is used in the synthesis of novel colchicine-derived nitrate esters as immunosuppressant agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-iodo-3-methylbenzoate is used in the synthesis of novel colchicine-derived nitrate esters as immunosuppressant agents.

Solubility
Slightly soluble in methanol.

Notes
Light sensitive. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5471-81-8
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD02683863
InChI Key HCSGWQGKCVQIRM-UHFFFAOYSA-N
Synonym 4-iodo-3-methylbenzoic acid methyl ester, methyl 4-iodo-3-methyl-benzoate, 4-iodo-m-toluic acid methyl ester, benzoic acid, 4-iodo-3-methyl-, methyl ester, methyl4-iodo-3-methylbenzoate, acmc-1anqa, methyl 4-iodo-m-toluate, methyl 3methyl-4-iodobenzoate, methyl 3-methyl-4-iodobenzoate, 4-iodo-m-toluic acid, methyl ester
PubChem CID 231757
IUPAC Name methyl 4-iodo-3-methylbenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)I

Specifications

Melting Point 59°C to 62°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in methanol.
Formula Weight 276.07
Percent Purity 98%
Chemical Name or Material Methyl 4-iodo-3-methylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.