missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodo-2-methoxybenzoate, 98+%

Product Code. p-7028293
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15443287

Brand: Thermo Scientific Alfa Aesar B25529.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 148490-97-5 | C9H9IO3 | 292.072 g/mol

Intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148490-97-5
Molecular Formula C9H9IO3
Molecular Weight (g/mol) 292.072
MDL Number MFCD06797865
InChI Key MCNOTXROWOGSGU-UHFFFAOYSA-N
Synonym benzoic acid,4-iodo-2-methoxy-, methyl ester, 4-iodo-2-methoxybenzoic acid methyl ester, 4-iodo-2-methoxy-benzoic acid methyl ester, pubchem4974, acmc-20adk9, methyl4-iodo-2-methoxybenzoate, methyl-4-iodo-2-methoxybenzoate, 2-methoxy-4-iodo-benzoic acid methyl ester
PubChem CID 22619468
IUPAC Name methyl 4-iodo-2-methoxybenzoate
SMILES COC1=C(C=CC(=C1)I)C(=O)OC

Specifications

Melting Point 44°C to 46°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 292.07
Percent Purity ≥98%
Chemical Name or Material Methyl 4-iodo-2-methoxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.