missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxyphenylacetate, 98+%

Product Code. 11490737
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490737 500 g 500g
11470737 25 g 25g
11480737 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490737 Supplier Thermo Scientific Alfa Aesar Supplier No. B22533.36
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14199-15-6 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14199-15-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002387
InChI Key XGDZEDRBLVIUMX-UHFFFAOYSA-N
Synonym methyl 4-hydroxyphenylacetate, methyl 2-4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methyl ester, benzeneacetic acid, 4-hydroxy-, methyl ester, methyl 4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methl ester, acetic acid, p-hydroxyphenyl-, methyl ester, p-hydroxyphenylacetic acid methyl ester, 4-hydroxybenzeneacetic acid methyl ester, 4-hydroxy-phenyl-acetic acid methyl ester
PubChem CID 518900
ChEBI CHEBI:68078
IUPAC Name methyl 2-(4-hydroxyphenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)O

Specifications

Melting Point 55°C to 58°C
Boiling Point 163°C (5 mmHg)
Quantity 500 g
Beilstein 2087538
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material Methyl 4-hydroxyphenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.