missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(hydroxymethyl)pyridine-2-carboxylate, 95%

Product Code. p-7031600
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448185 250 mg 250mg
15428185 1 g 1g
15438185 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448185 Supplier Thermo Scientific Alfa Aesar Supplier No. H32731.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 317335-15-2 | C8H9NO3 | 167.164 g/mol

Methyl 4-(hydroxymethyl)pyridine-2 is used as an intermediate in pharmaceuticals and also finds use in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-(hydroxymethyl)pyridine-2 is used as an intermediate in pharmaceuticals and also finds use in chemical research.

Notes
Store in a cool place. Incompatible with acids, bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317335-15-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD09834350
InChI Key WEIFQBALQLWZFZ-UHFFFAOYSA-N
Synonym methyl 4-hydroxymethyl picolinate, methyl 4-hydroxymethyl pyridine-2-carboxylate, methyl 4-hydroxymethyl-pyridine-2-carboxylate, 4-hydroxymethyl-2-pyridinecarboxylic acid methyl ester, 2-pyridinecarboxylic acid, 4-hydroxymethyl-, methyl ester, 2-pyridinecarboxylicacid,4-hydroxymethyl-,methylester 9ci, 2-methoxycarbonylpyridine-4-methanol, 4-hydroxymethyl-pyridine-2-carboxylic acid methyl, 4-hydroxymethylpyridin-2-carboxylic acid methyl ester, 4-hydroxymethyl pyridine-2-carboxylic acid methyl ester
PubChem CID 23093785
IUPAC Name methyl 4-(hydroxymethyl)pyridine-2-carboxylate
SMILES COC(=O)C1=NC=CC(=C1)CO

Specifications

Melting Point 74°C to 76°C
Quantity 250 mg
Formula Weight 167.16
Percent Purity 95%
Chemical Name or Material Methyl 4-(hydroxymethyl)pyridine-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.