missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, 99%, Thermo Scientific Chemicals

Product Code. 11383616
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383616 1 g 1g
11393616 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383616 Supplier Thermo Scientific Alfa Aesar Supplier No. L14622.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39493-62-4
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.191
MDL Number MFCD01075661
InChI Key SEBIQMPNDQLYOX-UHFFFAOYSA-N
Synonym methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-ene-1-carboxylate, methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-enecarboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-enecarboxylate, acmc-20akgd, methyl 4-hydroxy-6-methyl-2-oxo-cyclohex-3-en-1-oate, methyl 2-hydroxy-6-methyl-4-oxo-2-cyclohexene-1-carboxylate, 3-cyclohexene-1-carboxylicacid, 4-hydroxy-6-methyl-2-oxo-, methyl ester, 6-methyl-2-hydroxy-4-oxo-2-cyclohexene-1-carboxylic acid methyl ester
PubChem CID 3766543
IUPAC Name methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-ene-1-carboxylate
SMILES CC1CC(=O)C=C(C1C(=O)OC)O

Specifications

Melting Point 122°C to 124°C
Quantity 1 g
Beilstein 5258501
Formula Weight 184.19
Percent Purity 99%
Chemical Name or Material Methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.