missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxy-3-methoxycinnamate, 99%

Product Code. 11425277
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. Quantity unitSize
11425277 10 g 10g
11415277 2 g 2g
2 options
This item is not returnable. View return policy

Product Code. 11425277

Brand: Thermo Scientific Alfa Aesar B22657.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2309-07-1 | C16H25ClN2O | 296.84 g/mol

Methyl 4-hydroxy-3-methoxycinnamate has antioxidant activities that provide protection against inflammation and cancer. It has got the ability to cross cell membranes. It inhibits the release of pro-inflammatory cytokines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2309-07-1
MDL Number MFCD00017208

Specifications

Melting Point 62°C to 65°C
Molecular Formula C11H12O4
Quantity 10 g
Synonym methyl ferulate, methyl 3-4-hydroxy-3-methoxyphenyl acrylate, ferulic acid methyl ester, ferulic acid methylester, methyl 4-hydroxy-3-methoxycinnamate, cinnamic acid, 4-hydroxy-3-methoxy-, methyl ester, unii-y98bua66rx, y98bua66rx, methyl 2e-3-4-hydroxy-3-methoxyphenyl prop-2-enoate, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl-, methyl ester
Solubility Information Slightly soluble in water.
InChI Key AUJXJFHANFIVKH-GQCTYLIASA-N
SMILES COC1=C(C=CC(=C1)C=CC(=O)OC)O
IUPAC Name methyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Molecular Weight (g/mol) 208.21
PubChem CID 5357283
ChEBI CHEBI:67379
Formula Weight 208.22
Percent Purity 99%
Chemical Name or Material Methyl 4-hydroxy-3-methoxycinnamate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.