Learn More
Methyl 4-hydroxy-3-methoxycinnamate, 99%
Description
Methyl 4-hydroxy-3-methoxycinnamate has antioxidant activities that provide protection against inflammation and cancer. It has got the ability to cross cell membranes. It inhibits the release of pro-inflammatory cytokines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 4-hydroxy-3-methoxycinnamate has antioxidant activities that provide protection against inflammation and cancer. It has got the ability to cross cell membranes. It inhibits the release of pro-inflammatory cytokines.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 62°C to 65°C |
| Molecular Formula | C16H25ClN2O |
| Quantity | 2 g |
| Synonym | oxymetazoline hydrochloride, oxymetazoline hcl, afrazine, ocuclear, lliadine, nasivin, nasivine, neonabel, nostrilla |
| Solubility Information | Slightly soluble in water. |
| InChI Key | BEEDODBODQVSIM-UHFFFAOYSA-N |
| SMILES | CC1=CC(=C(C(=C1CC2=NCCN2)C)O)C(C)(C)C.Cl |
| IUPAC Name | 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol;hydrochloride |
| Molecular Weight (g/mol) | 296.84 |
| PubChem CID | 66259 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.