missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxy-3,5-dimethoxybenzoate, 98+%

Product Code. 11469196
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469196 25 g 25g
11438820 5 g 5g
11479196 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469196 Supplier Thermo Scientific Alfa Aesar Supplier No. A18054.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 884-35-5 | C10H12O5 | 212.201 g/mol

The scheme of laccase-catalysed iodide oxidation includes stadium of MS interaction with oxidized laccase with concomitant production of MS(ox). Thereaction of MS(ox) with iodide produced triiodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The scheme of laccase-catalysed iodide oxidation includes stadium of MS interaction with oxidized laccase with concomitant production of MS(ox). Thereaction of MS(ox) with iodide produced triiodide.

Solubility
Soluble in alcohol, ether, slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 884-35-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00017199
InChI Key ZMXJAEGJWHJMGX-UHFFFAOYSA-N
Synonym methyl syringate, methyl 3,5-dimethoxy-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3,5-dimethoxy-, methyl ester, syringic acid methyl ester, methyl 4-hydroxy-3,5-dimethoxy-benzoate, acmc-1bjig, ksc496o5n, syringic acid monomethyl ester, rarechem al bf 0102, timtec-bb sbb016976
PubChem CID 70164
ChEBI CHEBI:45820
IUPAC Name methyl 4-hydroxy-3,5-dimethoxybenzoate
SMILES COC1=CC(=CC(=C1O)OC)C(=O)OC

Specifications

Melting Point 102°C to 108°C
Quantity 25 g
Solubility Information Soluble in alcohol,ether,slightly soluble in water.
Formula Weight 212.2
Percent Purity ≥98%
Chemical Name or Material Methyl 4-hydroxy-3,5-dimethoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.