missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formylbenzoate dimethyl acetal, 95%, Thermo Scientific™

Product Code. 10626641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10626641 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10626641 Supplier Thermo Scientific Acros Supplier No. 424720010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 42228-16-0
Color Colorless to Yellow
Boiling Point 144°C (3.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C11H14O4
Linear Formula H3COOCC6H4CH(OCH3)2
Refractive Index 1.5065 to 1.5085
MDL Number MFCD00027368
Quantity 1 g
Synonym methyl 4-dimethoxymethyl benzoate, benzoic acid, 4-dimethoxymethyl-, methyl ester, methyl 4-formylbenzoate dimethylacetal, methyl-4-formylbenzoate dimethylacetal, 4-methoxycarbonyl benzaldehyde dimethyl acetal, methyl p-formylbenzoate dimethyl acetal, methyl-4-formylbenzoatedimethylacetal, methyl 4-dimethoxymethyl benzenecarboxylate, methyl-4-dimethoxymethyl-benzoate, 4-dimethoxymethyl benzoic acid methyl ester
Solubility Information Solubility in water: negligible
InChI Key YUTFBICAYRWLGQ-UHFFFAOYSA-N
SMILES COC(C1=CC=C(C=C1)C(=O)OC)OC
IUPAC Name methyl 4-(dimethoxymethyl)benzoate
Molecular Weight (g/mol) 210.23
PubChem CID 170620
Formula Weight 210.23
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material Methyl 4-formylbenzoate dimethyl acetal, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.