missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formylbenzoate, 99%

Product Code. 10042830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10042830 50 g Glass bottle 50g
10246470 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10042830 Supplier Thermo Scientific Acros Supplier No. 223750500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1571-08-0 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-08-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00006950
InChI Key FEIOASZZURHTHB-UHFFFAOYSA-N
Synonym methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate
PubChem CID 15294
IUPAC Name methyl 4-formylbenzoate
SMILES COC(=O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 57.0°C to 64.0°C
Color Cream to White or Green
Boiling Point 265.0°C
Flash Point 144°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HCOC6H4CO2CH3
Quantity 50 g
Solubility Information Solubility in water: insoluble
Formula Weight 164.16
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Methyl 4-formylbenzoate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-formylbenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Methyl 4-formylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.