missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 4-formyl-2,5-dimethylpyrrole-3-carboxylate, 96%

Product Code. 11335539
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335539 1 g 1g
11345539 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335539 Supplier Alfa Aesar™ Supplier No. H50477.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-91-1
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.191
MDL Number MFCD00203870
InChI Key OHVCRNQTGBXAPW-UHFFFAOYSA-N
Synonym methyl 4-formyl-2,5-dimethylpyrrole-3-carboxylate, methyl 2,5-dimethyl-4-formyl-1h-pyrrole-3-carboxylate, 1h-pyrrole-3-carboxylicacid, 4-formyl-2,5-dimethyl-, methyl ester, acmc-20akgf, methyl 4-formyl-2,5-dimethyl-1h-pyrrole-3 carboxylate, methyl 2,5-dimethyl-4-formylpyrrole-3-carboxylate, methyl 2,5-dimethyl-4-formyl-pyrrole-3-carboxylate, 2,5-dimethyl-4-methoxycarbonyl-1h-pyrrole-3-carboxaldehyde, 1h-pyrrole-3-carboxylic acid, 4-formyl-2,5-dimethyl-, methyl ester 9ci
PubChem CID 2798463
IUPAC Name methyl 4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
SMILES CC1=C(C(=C(N1)C)C(=O)OC)C=O

Specifications

Quantity 1 g
Formula Weight 181.19
Percent Purity 96%
Chemical Name or Material Methyl 4-formyl-2,5-dimethylpyrrole-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.