missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluorobenzoylacetate, 95%

Product Code. p-7028306
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11467226

Brand: Thermo Scientific Alfa Aesar B25546.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63131-29-3 | C10H9FO3 | 196.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63131-29-3
Molecular Formula C10H9FO3
Molecular Weight (g/mol) 196.177
MDL Number MFCD00000355
InChI Key HGLVYXXPRSNKQN-UHFFFAOYSA-N
Synonym methyl 4-fluorobenzoylacetate, methyl 3-4-fluorophenyl-3-oxopropanoate, methyl p-fluorobenzoylacetate, 4-fluorobenzoylacetic acid methyl ester, methyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid methyl ester, benzenepropanoic acid, 4-fluoro-beta-oxo-, methyl ester, benzenepropanoic acid, 4-fluoro-.beta.-oxo-, methyl ester, pubchem16721, acmc-209nd6
PubChem CID 579425
IUPAC Name methyl 3-(4-fluorophenyl)-3-oxopropanoate
SMILES COC(=O)CC(=O)C1=CC=C(C=C1)F

Specifications

Density 1.228
Boiling Point 91°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.521
Quantity 1 g
Formula Weight 196.18
Percent Purity 95%
Chemical Name or Material Methyl 4-fluorobenzoylacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.