missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluorobenzoate, 97%

Product Code. 11497054
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497054 25 g 25g
11487054 5 g 5g
11407064 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497054 Supplier Thermo Scientific Alfa Aesar Supplier No. B21504.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 403-33-8 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-33-8
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00017959
InChI Key MSEBQGULDWDIRW-UHFFFAOYSA-N
Synonym methyl p-fluorobenzoate, 4-fluorobenzoic acid methyl ester, methyl 4-fluorobenzoic acid, benzoic acid, 4-fluoro-, methyl ester, methyl4-fluorobenzoate, methyl-4-fluorobenzoate, 4-fluoro-benzoic acid methyl ester, pubchem3566, acmc-1aef0, asischem p32019
PubChem CID 67878
IUPAC Name methyl 4-fluorobenzoate
SMILES COC(=O)C1=CC=C(C=C1)F

Specifications

Density 1.192
Boiling Point 195°C to 197°C
Flash Point 77°C (170°F)
Odor Aromatic
Refractive Index 1.495
Quantity 25 g
Beilstein 2085925
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material Methyl 4-fluorobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.