missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluoro-3-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog Number 15468098
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
329-59-9
C8H6FNO4
199.14
MFCD00196161
CNJJSTPBUHAEFH-UHFFFAOYSA-N
methyl 4-fluoro-3-nitro-benzoate, 4-fluoro-3-nitrobenzoic acid methyl ester, 4-fluoro-3-nitro-benzoic acid methyl ester, benzoic acid, 4-fluoro-3-nitro-, methyl ester, pubchem2045, acmc-209hxf, methyl 4-fluoro-3 nitrobenzoate, 4-carbomethoxy-2-nitrofluorobenzene, methyl4-fluoro-3-nitrobenzoate
5219721
methyl 4-fluoro-3-nitrobenzoate
COC(=O)C1=CC(=C(F)C=C1)[N+]([O-])=O
This item is not returnable. View return policy
329-59-9
C8H6FNO4
199.14
MFCD00196161
CNJJSTPBUHAEFH-UHFFFAOYSA-N
methyl 4-fluoro-3-nitro-benzoate, 4-fluoro-3-nitrobenzoic acid methyl ester, 4-fluoro-3-nitro-benzoic acid methyl ester, benzoic acid, 4-fluoro-3-nitro-, methyl ester, pubchem2045, acmc-209hxf, methyl 4-fluoro-3 nitrobenzoate, 4-carbomethoxy-2-nitrofluorobenzene, methyl4-fluoro-3-nitrobenzoate
5219721
methyl 4-fluoro-3-nitrobenzoate
COC(=O)C1=CC(=C(F)C=C1)[N+]([O-])=O

Methyl 4-fluoro-3-nitrobenzoate is used to prepare dimethyl 3-nitro-3',4-oxydibenzoate by reacting with 3-hydroxy-benzoic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-fluoro-3-nitrobenzoate is used to prepare dimethyl 3-nitro-3′,4-oxydibenzoate by reacting with 3-hydroxy-benzoic acid methyl ester.

Solubility
Soluble in ethanol, ether and methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 56°C to 59°C
Boiling Point 109°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Solubility Information Soluble in ethanol,ether and methanol. Insoluble in water.
Formula Weight 199.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 4-fluoro-3-nitrobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'