missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluoro-3-nitrobenzoate, 98%

Product Code. p-7053832
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15468098

Brand: Thermo Scientific Alfa Aesar H61542.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 329-59-9 | C8H6FNO4 | 199.14 g/mol

Methyl 4-fluoro-3-nitrobenzoate is used to prepare dimethyl 3-nitro-3',4-oxydibenzoate by reacting with 3-hydroxy-benzoic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-fluoro-3-nitrobenzoate is used to prepare dimethyl 3-nitro-3′,4-oxydibenzoate by reacting with 3-hydroxy-benzoic acid methyl ester.

Solubility
Soluble in ethanol, ether and methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-59-9
Molecular Formula C8H6FNO4
Molecular Weight (g/mol) 199.14
MDL Number MFCD00196161
InChI Key CNJJSTPBUHAEFH-UHFFFAOYSA-N
Synonym methyl 4-fluoro-3-nitro-benzoate, 4-fluoro-3-nitrobenzoic acid methyl ester, 4-fluoro-3-nitro-benzoic acid methyl ester, benzoic acid, 4-fluoro-3-nitro-, methyl ester, pubchem2045, acmc-209hxf, methyl 4-fluoro-3 nitrobenzoate, 4-carbomethoxy-2-nitrofluorobenzene, methyl4-fluoro-3-nitrobenzoate
PubChem CID 5219721
IUPAC Name methyl 4-fluoro-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(F)C=C1)[N+]([O-])=O

Specifications

Melting Point 56°C to 59°C
Boiling Point 109°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Solubility Information Soluble in ethanol,ether and methanol. Insoluble in water.
Formula Weight 199.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 4-fluoro-3-nitrobenzoate
compliance-icons
Product Identifier
  • Methyl 4-fluoro-3-nitrobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 4-fluoro-3-nitrobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.