missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Cyanobenzoate, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Methyl 4-Cyanobenzoate |
| CAS | 1129-35-7 |
| Molecular Formula | C9H7NO2 |
| Quantity | 25 g |
| Synonym | 4-Cyanobenzoic Acid Methyl Ester, p-Cyanobenzoic Acid Methyl Ester, 4-(Methoxycarbonyl)benzonitrile, Methyl p-Cyanobenzoate, p-Cyanobenzoic Acid Methyl Ester |
| InChI Formula | InChI=1S/C9H7NO2/c1-12-9(11)8-4-2-7(6-10)3-5-8/h2-5H,1H3 |
| SMILES | COC(=O)c1ccc(cc1)C#N |
| IUPAC Name | methyl 4-cyanobenzoate |
| Molecular Weight (g/mol) | 161.16 |
| Formula Weight | 161.0477 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction