missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-[(chlorosulfonyl)methyl]benzoate, 95%, Thermo Scientific™

Product Code. 10257683
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10257683 5 g Amber Glass Bottle 5g
10506324 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10257683 Supplier Thermo Scientific Maybridge Supplier No. MO07948DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 130047-14-2
Molecular Formula C9H9ClO4S
Molecular Weight (g/mol) 248.68
MDL Number MFCD07778383
InChI Key VTQZPLOSFBIFGF-UHFFFAOYSA-N
Synonym methyl 4-chlorosulfonyl methyl benzoate, 4-methoxycarbonyl phenyl methanesulphonyl chloride, benzoic acid, 4-chlorosulfonyl methyl-, methyl ester, benzoic acid,4-chlorosulfonyl methyl-, methyl ester, acmc-20dqi1, methyl 4-chlorosulfonylmethyl benzoate, methyl 4-chlorosulphonyl methyl benzoate, 4-chlorosulfonylmethylbenzoic acid methyl ester, 4-methoxycarbonyl phenyl methanesulfonyl chloride
PubChem CID 21804301
IUPAC Name methyl 4-(chlorosulfonylmethyl)benzoate
SMILES COC(=O)C1=CC=C(CS(Cl)(=O)=O)C=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 248.69
Percent Purity 95%
Chemical Name or Material Methyl 4-[(chlorosulfonyl)methyl]benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.