missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(chloromethyl)benzoate, 97%, Thermo Scientific™

Product Code. 10784174 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10784174 5 g Glass bottle 5g
10688984 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10784174 Supplier Thermo Scientific Acros Supplier No. 391310050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 34040-64-7
MDL Number MFCD00511224
InChI Key SATDLKYRVXFXRE-UHFFFAOYSA-N
Synonym methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester
PubChem CID 36664
IUPAC Name methyl 4-(chloromethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)CCl

Specifications

Melting Point 37.0°C to 40.0°C
Color White
Boiling Point 160.0°C to 165.0°C (30.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C9H9ClO2
Linear Formula ClCH2C6H4C(O)OCH3
Quantity 5 g
Molecular Weight (g/mol) 184.619
Formula Weight 184.62
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material Methyl 4-(chloromethyl)benzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-(chloromethyl)benzoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.