missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(chloroformyl)butyrate, 98%

Product Code. 15209626 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15209626 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15209626 Supplier Thermo Scientific Acros Supplier No. 126540500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1501-26-4 | C6H9ClO3 | 164.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1501-26-4
Density 1.1910g/mL
Boiling Point 110.0°C (17.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Molecular Formula C6H9ClO3
Linear Formula ClCO(CH2)3CO2CH3
Refractive Index 1.4435 to 1.4455
MDL Number MFCD00000756
Quantity 50 g
Specific Gravity 1.191
Synonym methyl 4-chloroformyl butyrate, methyl 5-chloro-5-oxovalerate, methyl 4-chloroformylbutyrate, methyl5-chloro-5-oxopentanoate, 5-chloro-5-oxopentanoic acid methyl ester, methyl glutaryl chloride, pentanoic acid, 5-chloro-5-oxo-, methyl ester, 4-chlorocarbonyl-butyric acid methyl ester, zlchem 559, pubchem14033
InChI Key JCAZSWWHFJVFPP-UHFFFAOYSA-N
SMILES COC(=O)CCCC(Cl)=O
IUPAC Name methyl 5-chloro-5-oxopentanoate
Molecular Weight (g/mol) 164.59
PubChem CID 73916
Formula Weight 164.59
Percent Purity 98%
Chemical Name or Material Methyl 4-(chloroformyl)butyrate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-(chloroformyl)butyrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.