missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chlorobenzoate, Mikromol™

Product Code. 30051139
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051139 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051139 Supplier Mikromol Supplier No. MM0250.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 4-Chlorobenzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 4-Chlorobenzoate
CAS 1126-46-1
Analyte or Component Names Methyl 4-Chlorobenzoate
Shipping Condition Room Temperature
Molecular Formula C8 H7 Cl O2
Certifications/Compliance ISO 9001
Synonym Benzoic acid, 4-chloro-, methyl ester, Benzoic acid, p-chloro-, methyl ester (6CI,7CI,8CI), 4-(Methoxycarbonyl)chlorobenzene, 4-Carbomethoxyphenyl chloride, 4-Chlorobenzoic acid methyl ester, 5-Chloro-2-benzoic acid methyl ester, Methyl 4-chlorobenzoate, Methyl p-chlorobenzoate, NSC 8366, p-Chlorobenzoic acid methyl ester, Decitabine Related Compound D
InChI Formula InChI=1S/C8H7ClO2/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5H,1H3
SMILES COC(=O)c1ccc(Cl)cc1
IUPAC Name methyl 4-chlorobenzoate
Molecular Weight (g/mol) 170.593
Quantity 25 mg
Formula Weight 170.0135 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.