missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chloroacetoacetate, TRC

Product Code. 30468947
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468947 10 g 10g
30477547 1 g 1g
30484258 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30468947 Supplier Toronto Research Chemicals Supplier No. TRCM29455510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32807-28-6
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
Synonym 4-Chloro-3-oxobutanoic Acid Methyl Ester, 4-Chloroacetoacetic Acid Methyl Ester, 4-Chloro-3-oxo-butyric Acid Methyl Ester, Methyl (Chloroacetyl)acetate, Methyl 3-Oxo-4-chlorobutanoate, Methyl 4-Chloro-3-oxobutanoate, Methyl 4-chloro-3-oxobutyrate, Methyl gamma-Chloroacetoacetate, Methyl ω-Chloroacetoacetate
IUPAC Name methyl 4-chloro-3-oxobutanoate
SMILES COC(=O)CC(=O)CCl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H7ClO3/c1-9-5(8)2-4(7)3-6/h2-3H2,1H3
Formula Weight 150.0084
Chemical Name or Material Methyl 4-Chloroacetoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.