missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(bromomethyl)benzoate, 98%

Product Code. 11444280
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444280 5 g 5g
11454280 25 g 25g
11464280 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444280 Supplier Thermo Scientific Alfa Aesar Supplier No. A10557.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2417-72-3 | C9H9BrO2 | 229.07 g/mol

Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

Solubility
Lowly soluble in water, highly soluble in ethanol and ether, not soluble in halogenated organic solvents

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2417-72-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00032453
InChI Key NLWBJPPMPLPZIE-UHFFFAOYSA-N
PubChem CID 256687
IUPAC Name methyl 4-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 53°C to 58°C
Boiling Point 130°C to 135°C (2 mmHg)
Quantity 5 g
UN Number UN1759
Beilstein 1867272
Synonym methyl 4-bromomethyl benzoate, 4-bromomethylbenzoic acid methyl ester, methyl4-bromomethyl benzoate, benzoic acid, 4-bromomethyl-, methyl ester, 4-bromomethyl benzoic acid methyl ester, p-methoxycarbonyl benzyl bromide, methyl p-bromomethyl benzoate, 4-bromomethyl-benzoic acid methyl ester, 4-carbomethoxybenzyl bromide
Solubility Information Lowly soluble in water,highly soluble in ethanol and ether,not soluble in halogenated organic solvents
Formula Weight 229.07
Percent Purity 98%
Chemical Name or Material Methyl 4- (bromomethyl)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.