missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromocrotonate, tech. 85%

Product Code. 11477417
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477417 250 g 250g
11457417 10 g 10g
11467417 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477417 Supplier Thermo Scientific Alfa Aesar Supplier No. A15582.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1117-71-1 | C5H7BrO2 | 179.01 g/mol

It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1117-71-1
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.01
MDL Number MFCD00000246,MFCD09998308
InChI Key RWIKCBHOVNDESJ-NSCUHMNNSA-N
Synonym methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester
PubChem CID 5369175
IUPAC Name methyl (E)-4-bromobut-2-enoate
SMILES COC(=O)\C=C\CBr

Specifications

Density 1.524
Boiling Point 83°C to 85°C (13 mmHg)
Flash Point 91°C (195°F)
Linear Formula BrCH2CH=CHCO2CH3
Refractive Index 1.501
Quantity 250 g
UN Number UN1760
Beilstein 1745755
Solubility Information Slightly soluble in water.
Formula Weight 179.01
Percent Purity 85%
Grade Technical
Chemical Name or Material Methyl 4-bromocrotonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.