missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromobutyrate, 97%

Product Code. 11383276
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383276 50 g 50g
11373276 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383276 Supplier Thermo Scientific Alfa Aesar Supplier No. L12636.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4897-84-1 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4897-84-1
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00041482
InChI Key QAWFLJGZSZIZHO-UHFFFAOYSA-N
Synonym methyl 4-bromobutyrate, 4-bromobutyric acid methyl ester, methyl-4-bromobutyrate, butanoic acid, 4-bromo-, methyl ester, methyl4-bromobutyrate, butyric acid, 4-bromo-, methyl ester, methyl gamma-bromobutyrate, acmc-1amn9, 4-brombutyric acid methyl ester, 4-bromobutyric acid methyl-ester
PubChem CID 107604
IUPAC Name methyl 4-bromobutanoate
SMILES COC(=O)CCCBr

Specifications

Boiling Point 186°C to 187°C
Refractive Index 1.462
Quantity 50 g
Beilstein 1745618
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material Methyl 4-bromobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.