missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-bromo-3-hydroxythiophene-2-carboxylate, 97%
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 g
10 g
missing translation for 'unitSize'
10g
2.5g
Beschreibung
The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.
Solubility
Sparingly Soluble in water 0.95 g/L @ 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases, reducing agents.
The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.
Solubility
Sparingly Soluble in water 0.95 g/L @ 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases, reducing agents.
Spezifikation
Spezifikation
| Melting Point | 79°C to 80°C |
| Quantity | 2.5 g |
| Solubility Information | Sparingly Soluble in water 0.95g/L @ 25°C. |
| Formula Weight | 237.08 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl 4-bromo-3-hydroxythiophene-2-carboxylate |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur