missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromo-2-(bromomethyl)benzoate, 96%, Thermo Scientific™

Product Code. p-7034658
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459017 250 mg 250mg
15449017 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459017 Supplier Thermo Scientific Alfa Aesar Supplier No. H52350.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 4-bromo-2-(bromomethyl)benzoate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78471-43-9
Molecular Formula C9H8Br2O2
Molecular Weight (g/mol) 307.969
MDL Number MFCD04114319
InChI Key SGFACFBLUAWICV-UHFFFAOYSA-N
Synonym methyl 4-bromo-2-bromomethyl benzoate, methyl 4-bromo-2-bromomethyl-benzoate, 4-bromo-2-bromomethylbenzoic acid methyl ester, 4-bromo-2-bromomethyl-benzoic acid methyl ester, benzoic acid, 4-bromo-2-bromomethyl-, methyl ester, 4-bromo-2-bromomethyl benzoic acid methyl ester, acmc-209pec, methyl alpha,5-dibromo-o-toluate, methyl 4bromo-2-bromomethylbenzoate, methyl4-bromo-2-bromomethylbenzoate
PubChem CID 22031079
IUPAC Name methyl 4-bromo-2-(bromomethyl)benzoate
SMILES COC(=O)C1=C(C=C(C=C1)Br)CBr

Specifications

Melting Point 76°C to 78°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 307.97
Percent Purity 96%
Chemical Name or Material Methyl 4-bromo-2-(bromomethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.