missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-[beta-(5-Methylpyrazine-2-carboxamido)ethyl]benzene Sulfonamide Carbamate, TRC

Product Code. 30179876
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179876 500 mg 500mg
30180381 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179876 Supplier Toronto Research Chemicals Supplier No. TRCM322900500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33288-74-3
Molecular Formula C16 H18 N4 O5 S
Molecular Weight (g/mol) 378.4
Synonym Methyl [[4-[2-[[(5-Methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]carbamate, Methyl [[4-[2-[[(5-Methylpyrazin-2-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]carbamate, Carbamic acid, N-[[4-[2-[[(5-methyl-2-pyrazinyl)carbonyl]amino]ethyl]phenyl]sulfonyl]-, methyl ester, Carbamic acid, [[4-[2-[[(5-methylpyrazinyl)carbonyl]amino]ethyl]phenyl]sulfonyl]-, methyl ester (9CI), Carbamic acid, [[p-[2-(5-methylpyrazinecarboxamido)ethyl]phenyl]sulfonyl]-, methyl ester (8CI), N-[4-[β-(5-Methylpyrazine-2-carboxamido)ethyl]phenylsulfonyl]methylurethane
IUPAC Name methyl N-[4-[2-[(5-methylpyrazine-2-carbonyl)amino]ethyl]phenyl]sulfonylcarbamate
SMILES COC(=O)NS(=O)(=O)c1ccc(CCNC(=O)c2cnc(C)cn2)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H18N4O5S/c1-11-9-19-14(10-18-11)15(21)17-8-7-12-3-5-13(6-4-12)26(23,24)20-16(22)25-2/h3-6,9-10H,7-8H2,1-2H3,(H,17,21)(H,20,22)
Formula Weight 378.0998
Chemical Name or Material Methyl [(4-[2-[((5-Methylpyrazin-2-yl)carbonyl)amino]ethyl]phenyl)sulphonyl]carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.