missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Aminobenzoate, Mikromol™

Product Code. 17992196
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992196 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17992196 Supplier Mikromol Supplier No. MM1255.070025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 4-Aminobenzoate
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Methyl 4-Aminobenzoate
CAS 619-45-4
Analyte or Component Names Methyl 4-Aminobenzoate
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O2
Certifications/Compliance ISO 17025
Synonym 4-Aminobenzoic Acid Methyl Ester, Benzoic acid, p-amino-, methyl ester (6CI,7CI,8CI), (4-(Methoxycarbonyl)phenyl)amine, 4-(Carbomethoxy)aniline, 4-(Methoxycarbonyl)aniline, 4-Aminobenzenecarboxylic acid methyl ester, 4-Aminobenzoic acid methyl ester, Methyl 4-aminobenzoate, Methyl 4-aminophenylcarboxylate, Methyl aniline-4-carboxylate, Methyl p-aminobenzoate, NSC 3783, p-(Methoxycarbonyl)aniline, p-Aminobenzoic acid methyl ester, p-Carbomethoxyaniline, Benzocaine Impurity H
InChI Formula InChI=1S/C8H9NO2/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5H,9H2,1H3
SMILES COC(=O)c1ccc(N)cc1
IUPAC Name methyl 4-aminobenzoate
Molecular Weight (g/mol) 151.16
Quantity 25 mg
Formula Weight 151.0633 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.