missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-aminobenzoate, 98%

Product Code. 11419233
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11419233

Brand: Thermo Scientific Alfa Aesar A11715.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-45-4 | C8H9NO2 | 151.165 g/mol

Methyl 4-aminobenzoate is used in the synthesis of guanidine alkaloids, (±)-martinelline and (±)-martinellic acid by a protic acid catalyzed hetero Diels-Alder coupling reaction with N-Cbz 2-pyrroline. It is also used in organic synthesis and acts as a dye intermediate. Further, it is used to prepare 4-(2-methyl-4-oxo-4H-quinazolin-3-yl)-benzoic acid methyl ester by reacting with 2-acetylamino-benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-aminobenzoate is used in the synthesis of guanidine alkaloids, (±)-martinelline and (±)-martinellic acid by a protic acid catalyzed hetero Diels-Alder coupling reaction with N-Cbz 2-pyrroline. It is also used in organic synthesis and acts as a dye intermediate. Further, it is used to prepare 4-(2-methyl-4-oxo-4H-quinazolin-3-yl)-benzoic acid methyl ester by reacting with 2-acetylamino-benzoic acid.

Solubility
Soluble in alcohol and ether. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-45-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007891
InChI Key LZXXNPOYQCLXRS-UHFFFAOYSA-N
Synonym 4-aminobenzoic acid methyl ester, methyl p-aminobenzoate, benzoic acid, 4-amino-, methyl ester, p-methoxycarbonyl aniline, 4-methoxycarbonyl aniline, p-aminobenzoic acid methyl ester, methyl aniline-4-carboxylate, 4-carbomethoxyaniline, methyl4-aminobenzoate, benzoic acid, p-amino-, methyl ester
PubChem CID 12082
IUPAC Name methyl 4-aminobenzoate
SMILES COC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 110°C to 114°C
Assay Percent Range 98%
Quantity 25 g
Beilstein 775913
Solubility Information Soluble in alcohol and ether. Slightly soluble in water.
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material Methyl 4-aminobenzoate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.