missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-aminobenzoate, 98%

Product Code. 11439233
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439233 500 g 500g
11419233 25 g 25g
11429233 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439233 Supplier Thermo Scientific Alfa Aesar Supplier No. A11715.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-45-4 | C8H9NO2 | 151.165 g/mol

Methyl 4-aminobenzoate is used in the synthesis of guanidine alkaloids, (±)-martinelline and (±)-martinellic acid by a protic acid catalyzed hetero Diels-Alder coupling reaction with N-Cbz 2-pyrroline. It is also used in organic synthesis and acts as a dye intermediate. Further, it is used to prepare 4-(2-methyl-4-oxo-4H-quinazolin-3-yl)-benzoic acid methyl ester by reacting with 2-acetylamino-benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-aminobenzoate is used in the synthesis of guanidine alkaloids, (±)-martinelline and (±)-martinellic acid by a protic acid catalyzed hetero Diels-Alder coupling reaction with N-Cbz 2-pyrroline. It is also used in organic synthesis and acts as a dye intermediate. Further, it is used to prepare 4-(2-methyl-4-oxo-4H-quinazolin-3-yl)-benzoic acid methyl ester by reacting with 2-acetylamino-benzoic acid.

Solubility
Soluble in alcohol and ether. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-45-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007891
InChI Key LZXXNPOYQCLXRS-UHFFFAOYSA-N
Synonym 4-aminobenzoic acid methyl ester, methyl p-aminobenzoate, benzoic acid, 4-amino-, methyl ester, p-methoxycarbonyl aniline, 4-methoxycarbonyl aniline, p-aminobenzoic acid methyl ester, methyl aniline-4-carboxylate, 4-carbomethoxyaniline, methyl4-aminobenzoate, benzoic acid, p-amino-, methyl ester
PubChem CID 12082
IUPAC Name methyl 4-aminobenzoate
SMILES COC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 110°C to 114°C
Assay Percent Range 98%
Quantity 500 g
Beilstein 775913
Solubility Information Soluble in alcohol and ether. Slightly soluble in water.
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material Methyl 4-aminobenzoate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.