missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-nitrobenzoate, 97%

Product Code. 15437287
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437287 1 g 1g
15447287 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437287 Supplier Thermo Scientific Alfa Aesar Supplier No. H34348.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3987-92-6 | C8H8N2O4 | 196.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3987-92-6
Molecular Formula C8H8N2O4
Molecular Weight (g/mol) 196.162
MDL Number MFCD00017562
InChI Key HNTLUEZVPLRQEV-UHFFFAOYSA-N
Synonym methyl 4-amino-3-nitrobenzenecarboxylate, benzoic acid, 4-amino-3-nitro-, methyl ester, 4-amino-3-nitro-benzoic acid methyl ester, 4-methoxycarbonyl-2-nitroaniline, 4-amino-3-nitromethylbenzoate, methyl 3-nitro-4-aminobenzoate, 2-amino-5-methoxycarbonyl nitrobenzene, 3-nitro-4-aminobenzoic acid methyl ester, 4-amino-3-nitrobenzoicacidmethylester, acmc-209j8j
PubChem CID 3941008
IUPAC Name methyl 4-amino-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)N)[N+](=O)[O-]

Specifications

Melting Point 188°C to 191°C
Quantity 1 g
Formula Weight 196.16
Percent Purity 97%
Chemical Name or Material Methyl 4-amino-3-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.