missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-chlorobenzoate, 97%, Thermo Scientific Chemicals

Catalog Number 10195353
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
84228-44-4
C8H8ClNO2
185.61
MFCD01320686
WKVJBYKFGNVWLM-UHFFFAOYSA-N
4-amino-3-chlorobenzoic acid methyl ester, 4-amino-3-chloro-benzoic acid methyl ester, methyl 4-amino-3-chloro-benzoate, benzoic acid, 4-amino-3-chloro-, methyl ester, 2-chloro-4-methoxycarbonyl aniline, pubchem10804, akos bb-3098, timtec-bb sbb003712, acmc-20a51m, methyl-4-amino-3-chlorobenzoate
1515284
methyl 4-amino-3-chlorobenzoate
COC(=O)C1=CC(=C(C=C1)N)Cl
This item is not returnable. View return policy
84228-44-4
C8H8ClNO2
185.61
MFCD01320686
WKVJBYKFGNVWLM-UHFFFAOYSA-N
4-amino-3-chlorobenzoic acid methyl ester, 4-amino-3-chloro-benzoic acid methyl ester, methyl 4-amino-3-chloro-benzoate, benzoic acid, 4-amino-3-chloro-, methyl ester, 2-chloro-4-methoxycarbonyl aniline, pubchem10804, akos bb-3098, timtec-bb sbb003712, acmc-20a51m, methyl-4-amino-3-chlorobenzoate
1515284
methyl 4-amino-3-chlorobenzoate
COC(=O)C1=CC(=C(C=C1)N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 106°C to 110°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula NH2C6H3(Cl)COOH
Quantity 1 g
Formula Weight 185.61
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-amino-3-chlorobenzoate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-amino-3-chlorobenzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'