missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-bromo-5-nitrobenzoate, 97%, Thermo Scientific Chemicals

Product Code. 15406027
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406027 5 g 5g
15496017 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406027 Supplier Thermo Scientific Alfa Aesar Supplier No. H33216.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105655-17-2 | Molecular Formula: C8H7BrN2O4 | Molecular Weight: 275.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105655-17-2
Molecular Formula C8H7BrN2O4
Molecular Weight (g/mol) 275.058
MDL Number MFCD08443938
InChI Key PRTLMEWWYBJZPN-UHFFFAOYSA-N
Synonym methyl 4-amino-3-bromo-5-nitrobenzenecarboxylate, benzoic acid, 4-amino-3-bromo-5-nitro-, methyl ester, acmc-20aq1i, methylaminobromonitrobenzenecarboxylate, 3-nitro-4-amino-5-bromobenzoic acid methyl ester, benzoicacid,4-amino-3-bromo-5-nitro-,methylester, 4-amino-3-bromo-5-nitro-benzoic acid methyl ester
PubChem CID 13685347
IUPAC Name methyl 4-amino-3-bromo-5-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)Br)N)[N+](=O)[O-]

Specifications

Melting Point 107°C to 109°C
Quantity 5 g
Formula Weight 275.06
Percent Purity 97%
Chemical Name or Material Methyl 4-amino-3-bromo-5-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.