missing translation for 'onlineSavingsMsg'
Learn More

methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride, Thermo Scientific™

Product Code. 10249412
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10249412 1 g 1g
10635155 5 g 5g
10487174 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10249412 Supplier Thermo Scientific Maybridge Supplier No. MO00998DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 180258-45-1
Molecular Formula C7H11ClN2O2
Molecular Weight (g/mol) 190.63
MDL Number MFCD01318761
InChI Key HEOKCJUUKIPIMM-UHFFFAOYSA-N
Synonym methyl 4-amino-1-methyl-1h-pyrrole-2-carboxylate hydrochloride, methyl 4-amino-1-methylpyrrole-2-carboxylate hydrochloride, methyl-4-amino-1-methyl pyrrole-2-carboxylate hydrochloride, 4-amino-1-methyl-1h-pyrrole-2-carboxylic acid-methyl ester hcl, 4-amino-1-methylpyrrole-2-carboxylic acid methyl ester hydrochloride, h-nh 4-mepyl-2-ome hcl, methyl 4-amino-1-methyl-pyrrole-2-carboxylate hydrochloride, methyl-4-amino-1-methyl pyrrole-2-carboxylate, hcl, n-methyl-2-carbomethoxy-4-aminopyrrole hydrochloride, 3-amino-1-methyl-5-methyloxycarbonylpyrrol hydrochloride
PubChem CID 2794656
SMILES Cl.COC(=O)C1=CC(N)=CN1C

Specifications

Assay Percent Range 95%
Quantity 1 g
IUPAC Name methyl 4-amino-1-methylpyrrole-2-carboxylate;hydrochloride
Formula Weight 190.63
Percent Purity 95%
Chemical Name or Material Methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.