missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(Acetylamino)-5-chloro-2-methoxybenzoate, Mikromol™

Product Code. 30050572
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050572 25 mg 25mg
30052174 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050572 Supplier Mikromol Supplier No. MM0073.040025
 Restricted Item
Estimated Shipment: 20-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 4-(Acetylamino)-5-chloro-2-methoxybenzoate
TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Methyl 4-(Acetylamino)-5-chloro-2-methoxybenzoate
CAS 4093-31-6
Analyte or Component Names Methyl 4-(Acetylamino)-5-chloro-2-methoxybenzoate
Shipping Condition Room Temperature
Molecular Formula C11 H12 Cl N O4
Certifications/Compliance ISO 17025
Synonym Metoclopramide Imp. B (EP), Metoclopramide USP Related Compound B, Metoclopramide USP RC B, Methyl 4-(Acetylamino)-5-chloro-2-methoxybenzoate, Metoclopramide Related Compound B, Metoclopramide Hydrochloride Monohydrate Impurity B, Metoclopramide Impurity B
InChI Formula InChI=1S/C11H12ClNO4/c1-6(14)13-9-5-10(16-2)7(4-8(9)12)11(15)17-3/h4-5H,1-3H3,(H,13,14)
SMILES COC(=O)c1cc(Cl)c(NC(=O)C)cc1OC
IUPAC Name methyl 4-acetamido-5-chloro-2-methoxybenzoate
Molecular Weight (g/mol) 257.67
Quantity 25 mg
Formula Weight 257.0455 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.