missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Acetamido-2-methoxybenzoate, TRC

Product Code. 30135832
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135832 25 g 25g
30009209 5 g 5g
17970585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135832 Supplier Toronto Research Chemicals Supplier No. TRCM27522025G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4093-29-2
Molecular Formula C11 H13 N O4
Molecular Weight (g/mol) 223.23
Synonym Methyl 4-(Acetylamino)-2-methoxybenzoate, Metoclopramide Imp. D (EP), Metoclopramide Imp. D (EP), Metoclopramide USP Related Compound D, Metoclopramide USP RC D, Methyl 4-(Acetylamino)-2-methoxybenzoate, Metoclopramide Related Compound D, Metoclopramide Hydrochloride Monohydrate Impurity D, Metoclopramide Impurity D
IUPAC Name methyl 4-acetamido-2-methoxybenzoate
SMILES COC(=O)c1ccc(NC(=O)C)cc1OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C11H13NO4/c1-7(13)12-8-4-5-9(11(14)16-3)10(6-8)15-2/h4-6H,1-3H3,(H,12,13)
Formula Weight 223.0845
Chemical Name or Material Methyl 4-(Acetylamino)-2-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.