missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Acetamido-2-hydroxybenzoate, TRC

Product Code. 30138474
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138474 10 g 10g
17958874 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138474 Supplier Toronto Research Chemicals Supplier No. TRCM27528010G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4093-28-1
Molecular Formula C10 H11 N O4
Molecular Weight (g/mol) 209.2
Synonym Methyl 4-(Acetylamino)-2-hydroxybenzoate
IUPAC Name methyl 4-acetamido-2-hydroxybenzoate
SMILES COC(=O)c1ccc(NC(=O)C)cc1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H11NO4/c1-6(12)11-7-3-4-8(9(13)5-7)10(14)15-2/h3-5,13H,1-2H3,(H,11,12)
Formula Weight 209.0688
Chemical Name or Material Methyl 4-(Acetylamino)-2-hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.