missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-(5-formyl-2-thienyl)benzoate, 95%, Thermo Scientific™
£29.90 - £115.00
Chemical Identifiers
CAS | 639816-37-8 |
---|---|
Molecular Formula | C13H10O3S |
Molecular Weight (g/mol) | 246.28 |
MDL Number | MFCD06200909 |
InChI Key | YWZJKUTYQOHLGQ-UHFFFAOYSA-N |
Synonym | methyl 4-5-formylthiophen-2-yl benzoate, methyl 4-5-formyl-2-thienyl benzoate, methyl 4-5-formyl-2-thienyl benzenecarboxylate, 4-5-formyl-thiophen-2-yl-benzoic acid methyl ester, amtda036, methylformylthienylbenzenecarboxylate, 4-5-formylthiophen-2-yl benzoic acid methyl ester, 4-5-formyl-thiophene-2-yl-benzoic acid methyl ester |
PubChem CID | 2763348 |
IUPAC Name | methyl 4-(5-formylthiophen-2-yl)benzoate |
SMILES | COC(=O)C1=CC=C(C=C1)C1=CC=C(S1)C=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
639816-37-8 | |
246.28 | |
YWZJKUTYQOHLGQ-UHFFFAOYSA-N | |
2763348 | |
COC(=O)C1=CC=C(C=C1)C1=CC=C(S1)C=O |
C13H10O3S | |
MFCD06200909 | |
methyl 4-5-formylthiophen-2-yl benzoate, methyl 4-5-formyl-2-thienyl benzoate, methyl 4-5-formyl-2-thienyl benzenecarboxylate, 4-5-formyl-thiophen-2-yl-benzoic acid methyl ester, amtda036, methylformylthienylbenzenecarboxylate, 4-5-formylthiophen-2-yl benzoic acid methyl ester, 4-5-formyl-thiophene-2-yl-benzoic acid methyl ester | |
methyl 4-(5-formylthiophen-2-yl)benzoate |
Specifications
639816-37-8 | |
MFCD06200909 | |
YWZJKUTYQOHLGQ-UHFFFAOYSA-N | |
methyl 4-(5-formylthiophen-2-yl)benzoate | |
2763348 | |
95% |
C13H10O3S | |
methyl 4-5-formylthiophen-2-yl benzoate, methyl 4-5-formyl-2-thienyl benzoate, methyl 4-5-formyl-2-thienyl benzenecarboxylate, 4-5-formyl-thiophen-2-yl-benzoic acid methyl ester, amtda036, methylformylthienylbenzenecarboxylate, 4-5-formylthiophen-2-yl benzoic acid methyl ester, 4-5-formyl-thiophene-2-yl-benzoic acid methyl ester | |
COC(=O)C1=CC=C(C=C1)C1=CC=C(S1)C=O | |
246.28 | |
246.29 | |
Methyl 4-(5-formyl-2-thienyl)benzoate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures