missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-dimethoxy-2-nitrobenzoate, 97%

Product Code. 11478810
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
500 g
Unit Size:
10g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478810 50 g 50g
11468810 10 g 10g
11488810 250 g 250g
11445233 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11478810 Supplier Thermo Scientific Alfa Aesar Supplier No. A12836.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 26791-93-5 | C10H11NO6 | 241.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26791-93-5
Molecular Formula C10H11NO6
Molecular Weight (g/mol) 241.199
MDL Number MFCD00007239
InChI Key SYYKLKHBZGFKOC-UHFFFAOYSA-N
Synonym methyl 6-nitroveratrate, benzoic acid, 4,5-dimethoxy-2-nitro-, methyl ester, 6-nitroveratric acid methyl ester, 4,5-dimethoxy-2-nitrobenzoic acid, methyl ester, methyl 4,5-dimethoxy-2-nitro-benzoate, acmc-1cptq, ksc496m9r, methyl 3,4-dimethoxy-6-nitrobenzoate, methyl 3,4-dimethoxy-6-nitro-benzoate, methyl 4,5-dimethoxy-2-nitrobenzoate
PubChem CID 117867
IUPAC Name methyl 4,5-dimethoxy-2-nitrobenzoate
SMILES COC1=C(C=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC

Specifications

Melting Point 142°C to 144°C
Quantity 50 g
Beilstein 2381747
Formula Weight 241.2
Percent Purity 97%
Chemical Name or Material Methyl 4,5-dimethoxy-2-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.