missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 11415676
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415676 5 g 5g
11405676 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415676 Supplier Thermo Scientific Alfa Aesar Supplier No. B20780.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 96232-71-2 | Molecular Formula: C6H4Br2O3S | Molecular Weight: 315.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96232-71-2
Molecular Formula C6H4Br2O3S
Molecular Weight (g/mol) 315.96
MDL Number MFCD00052082
InChI Key YZPNGCXFJCPJSV-XQRVVYSFSA-N
Synonym methyl4,5-dibromo-3-hydroxythiophene-2-carboxylate, 4,5-dibromo-3-hydroxythiophene-2-carboxylic acid methyl ester, 2z-4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one, acmc-20akgp, maybridge1_003706, methyl 4,5-dibromo-3-hydroxy-2-thiophenecarboxylate, 4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one, methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylat, 2-thiophenecarboxylicacid,4,5-dibromo-3-hydroxy-,methyl ester, 2e-4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one
PubChem CID 2777613
IUPAC Name methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate
SMILES CO\C(O)=C1/SC(Br)=C(Br)C1=O

Specifications

Melting Point 125°C to 128°C
Quantity 5 g
Formula Weight 316.89
Percent Purity 97%
Chemical Name or Material Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.