missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 11326875
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11326875 25 g Amber Glass Bottle 25g
10249852 1 g Amber Glass Bottle 1g
11935354 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326875 Supplier Thermo Scientific Maybridge Supplier No. GK01947EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 96232-71-2
Molecular Formula C6H4Br2O3S
Molecular Weight (g/mol) 315.96
MDL Number MFCD00052082
InChI Key YZPNGCXFJCPJSV-XQRVVYSFSA-N
Synonym methyl4,5-dibromo-3-hydroxythiophene-2-carboxylate, 4,5-dibromo-3-hydroxythiophene-2-carboxylic acid methyl ester, 2z-4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one, acmc-20akgp, maybridge1_003706, methyl 4,5-dibromo-3-hydroxy-2-thiophenecarboxylate, 4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one, methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylat, 2-thiophenecarboxylicacid,4,5-dibromo-3-hydroxy-,methyl ester, 2e-4,5-dibromo-2-hydroxy methoxy methylidene thiophen-3-one
PubChem CID 2777613
IUPAC Name methyl 4,5-dibromo-3-hydroxythiophene-2-carboxylate
SMILES CO\C(O)=C1/SC(Br)=C(Br)C1=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 315.97
Percent Purity 97%
Chemical Name or Material Methyl 4, 5-dibromo-3-hydroxythiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.