missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,4-dimethyl-3-oxovalerate, 95%

Product Code. 11332326
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332326 5 g 5g
11342326 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332326 Supplier Thermo Scientific Alfa Aesar Supplier No. L06007.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55107-14-7 | C8H14O3 | 158.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55107-14-7
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00008847
InChI Key XTXCFTMJPRXBBC-UHFFFAOYSA-N
Synonym methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2
PubChem CID 99597
IUPAC Name methyl 4,4-dimethyl-3-oxopentanoate
SMILES CC(C)(C)C(=O)CC(=O)OC

Specifications

Density 0.99
Boiling Point 67°C to 70°C (13 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.432
Quantity 5 g
Beilstein 1764796
Formula Weight 158.2
Percent Purity 95%
Chemical Name or Material Methyl 4,4-dimethyl-3-oxovalerate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.