missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,4,4-trifluoroacetoacetate, 95%

Product Code. 11348255
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348255 5 g 5g
11358255 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348255 Supplier Thermo Scientific Alfa Aesar Supplier No. L10140.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83643-84-9 | C5H5F3O3 | 170.087 g/mol

It is used as a drug intermediate. It is a cyclization, oxidation and halogenating reaction catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a drug intermediate. It is a cyclization, oxidation and halogenating reaction catalyst.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83643-84-9
Molecular Formula C5H5F3O3
Molecular Weight (g/mol) 170.087
MDL Number MFCD00041004
InChI Key LKMUBWWZTSZGGV-UHFFFAOYSA-N
PubChem CID 550266
IUPAC Name methyl 4,4,4-trifluoro-3-oxobutanoate
SMILES COC(=O)CC(=O)C(F)(F)F

Specifications

Density 1.34
Boiling Point 117°C to 118°C
Flash Point 40°C (104°F)
Refractive Index 1.372
Quantity 5 g
UN Number UN3272
Beilstein 1777343
Synonym methyl trifluoroacetoacetate, methyl 4,4,4-trifluoroacetoacetate, 4,4,4-trifluoroacetoacetic acid methyl ester, methyl-4,4,4-trifluoro-3-oxobutanoate, butanoic acid, 4,4,4-trifluoro-3-oxo-, methyl ester, acmc-209psv, methyltri-fluoroacetoacetate, methyl4,4,4-trifluoroacetoacetate, methyl-4,4,4-trifluoroacetoacetate, methyl 3-oxo-4,4,4-trifluorobutyrate
Formula Weight 170.09
Percent Purity 95%
Chemical Name or Material Methyl 4,4,4-trifluoroacetoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.