missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl [[4-[2-[(5-Chloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulphonyl]carbamate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Methyl [[4-[2-[(5-Chloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulphonyl]carbamate
Specifications
Specifications
| Chemical Name or Material | Methyl [(4-[2-[(5-Chloro-2-methoxybenzoyl)amino]ethyl]phenyl)sulfonyl]carbamate |
| CAS | 21165-77-5 |
| Analyte or Component Names | Methyl [(4-[2-[(5-Chloro-2-methoxybenzoyl)amino]ethyl]phenyl)sulfonyl]carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H19 Cl N2 O6 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Glibenclamide Imp. B (EP), Glyburide USP Related Compound B (AS), Glyburide USP RC B (AS), Methyl [[4-[2-[(5-Chloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulphonyl]carbamate, Glyburide Related Compound B, Glibenclamide Impurity B |
| InChI Formula | InChI=1S/C18H19ClN2O6S/c1-26-16-8-5-13(19)11-15(16)17(22)20-10-9-12-3-6-14(7-4-12)28(24,25)21-18(23)27-2/h3-8,11H,9-10H2,1-2H3,(H,20,22)(H,21,23) |
| SMILES | COC(=O)NS(=O)(=O)c1ccc(CCNC(=O)c2cc(Cl)ccc2OC)cc1 |
| IUPAC Name | methyl N-[4-[2-[(5-chloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulfonylcarbamate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction